Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR114 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102082 Supplier Invitrogen™ Supplier No. PA5102082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR114.

This gene encodes a member of the adhesion family of G-protein coupled receptors. Members of this family are characterized by long N-termini and multiple functional domains. They may play a role in the immune system as well as in the central nervous system. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR114
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRG5
Gene Accession No. Q8IZF4
Gene Alias ADGRG5; adhesion G protein-coupled receptor G5; adhesion G-protein coupled receptor G5; G protein-coupled receptor 114; G protein-coupled receptor PGR27; GPR114; G-protein coupled receptor 114; G-protein coupled receptor PGR27; PGR27; probable G-protein coupled receptor 114; UNQ2524/PRO6017
Gene Symbols ADGRG5
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADGRG5(Accession Q8IZF4), corresponding to amino acid residues V190-V240.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221188
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.