Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR115 Antibody (527018), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB54371H
Description
GPR115 Monoclonal antibody specifically detects GPR115 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
GPR115 | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
527018 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
FLJ38076, G protein-coupled receptor 115, G-protein coupled receptor PGR18, PGR18G-protein coupled receptor 115, probable G-protein coupled receptor 115, seven transmembrane helix receptor | |
Chinese hamster ovary cell line CHO-derived recombinant human GPR115, Ser22-Ala347, Accession # Q8IZF3 | |
0.1 mL | |
GPCR | |
221393 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction