Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR115 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GPR115 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GPR115 Polyclonal specifically detects GPR115 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GPR115 | |
Polyclonal | |
Rabbit | |
GPCR | |
FLJ38076, G protein-coupled receptor 115, G-protein coupled receptor PGR18, PGR18G-protein coupled receptor 115, probable G-protein coupled receptor 115, seven transmembrane helix receptor | |
ADGRF4 | |
IgG | |
Affinity Purified |
Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Unconjugated | |
RUO | |
Human | |
221393 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QSPEGKPKTGRIQEKCEGPCISSSNCSQPCAKDFHGEIGFTCNQKKWQKSAETCTSLSVEKLFKDSTGASRLSVAAPSIPLHILDFRAPETIESVAQGIRKNCPFDYACITDMVKSSETTSGNIAFIVELLKNISTDLSDNVTR | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title