Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR115 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102083 Supplier Invitrogen™ Supplier No. PA5102083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR115.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR115 is a member of this family (subfamily Orphan-B). ESTs have been isolated from breast, heart, and placenta libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR115
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRF4
Gene Accession No. Q8IZF3, Q9D2L6
Gene Alias 4632435A09Rik; ADGRF4; Adhesion G protein-coupled receptor F4; AV239010; FLJ38076; G protein-coupled receptor 115; GPR115; G-protein coupled receptor 115; G-protein coupled receptor PGR18; PGR18; probable G-protein coupled receptor 115; RGD1565216; seven transmembrane helix receptor
Gene Symbols ADGRF4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADGRF4(Accession Q8IZF3), corresponding to amino acid residues R660-G695.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221393, 78249
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.