Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR116 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314343
This item is not returnable.
View return policy
Description
GPR116 Polyclonal antibody specifically detects GPR116 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GPR116 | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ90640, G protein-coupled receptor 116, Ig-Hepta homolog, KIAA0758DKFZp564O1923, KPG_001, probable G-protein coupled receptor 116 | |
Synthetic 20 amino acid peptide from internal region of human GPR116. | |
50 μg | |
GPCR | |
221395 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction