Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR119 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533615
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bat (100%); Marmoset, Mouse, Hamster, Panda, Dog, Horse (95%); Rat, Elephant, Rabbit (90%); Bovine, Pig (80%). Proteinase K treatment is required for antigen retrieval.
GPCR GPR119 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.
Specifications
GPR119 | |
Polyclonal | |
Unconjugated | |
GPR119 | |
G protein-coupled receptor 119; glucose-dependent insulinotropic receptor; GPCR2; Gpr119; G-protein coupled receptor 119; G-protein coupled receptor 2; HGPCR2; RP1-20I3.4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
139760 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8TDV5 | |
GPR119 | |
Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human GPR119. | |
50 μg | |
Primary | |
Human, Bat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction