Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR119 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575326
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
GPCR GPR119 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.
Specifications
GPR119 | |
Polyclonal | |
Unconjugated | |
GPR119 | |
G protein-coupled receptor 119; glucose-dependent insulinotropic receptor; GPCR2; Gpr119; G-protein coupled receptor 119; G-protein coupled receptor 2; HGPCR2; RP1-20I3.4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
139760, 236781, 302813 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7TQN8, Q7TQP3, Q8TDV5 | |
GPR119 | |
Synthetic peptide corresponding to amino acids 190-240 of Human GPR119. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction