Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR12 Antibody (584940), Biotin, Novus Biologicals™
SDP

Catalog No. NB375894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375894 Supplier Novus Biologicals Supplier No. FAB6738B
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR12 Monoclonal antibody specifically detects GPR12 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry

Specifications

Antigen GPR12
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 584940
Conjugate Biotin
Dilution Flow Cytometry, Immunohistochemistry
Formulation PBS
Gene Alias FLJ18149, FLJ97704, G protein-coupled receptor 12, GPCR12, GPCR21, G-protein coupled receptor 12, MGC138349, MGC138351
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR12, Met1-Val334, Accession # P47775
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2835
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.