Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532793
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532793 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532793 Supplier Invitrogen™ Supplier No. PA532793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bovine, Hamster, Panda, Horse, Pig, Opossum (100%); Bat, Elephant, Rabbit (94%); Turkey, Chicken (88%); Platypus, Xenopus (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR12 is an orphan G-protein-coupled receptor which constitutively activates the Gs protein and increases intracellular cAMP. GPR12 can also be activated by sphingosylphosphorylcholine to promote various cellular functions which include proliferation, neurite outgrowth, cell survival and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR12
Gene Accession No. P30951, P35412, P47775
Gene Alias FLJ18149; FLJ97704; G protein-coupled receptor 12; GPCR01; GPCR12; Gpcr20; Gpcr21; Gpr12; G-protein coupled receptor 12; MGC138349; MGC138351; PPP1R84; protein phosphatase 1, regulatory subunit 84; R334
Gene Symbols GPR12
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 1st extracellular domain of human GPR12 / GPCR12.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050970, 100736641, 100764513, 14738, 2835, 486034, 522747, 80840
Target Species Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.