Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR123 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560114
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560114 Supplier Invitrogen™ Supplier No. PA560114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PCKMTNLQAA QGHASCLSPA TPCCAKMHCE PLTADEAHVH LQEEGAFGHD PHLHGCLQGR TKPPYFSRHP AEEPEYAYHI PSSLDGSPRS SRTDSPPSSL DG Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 68%.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR123 (G protein-coupled receptor 123) is a 1,279 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 2 family and LN-TM7 subfamily. Existing as two alternatively spliced isoforms, GPR123 functions as an orphan receptor that is expressed in adult and fetal brain, and in adult spinal cord. The gene encoding GPR123 maps to human chromosome 10q26.3 and mouse chromosome 7 F4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR123
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADGRA1
Gene Accession No. Q86SQ6
Gene Alias 2900059M17Rik; Adgra1; Adhesion G protein-coupled receptor A1; D7Ertd680e; G protein-coupled receptor 123; GPR123; G-protein coupled receptor 123; KIAA1828; probable G-protein coupled receptor 123
Gene Symbols ADGRA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR123. Recombinant protein control fragment (Product #RP-99985).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.