Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR123 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | GPR123 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2684
![]() ![]() |
Proteintech Group Inc
200751AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to GPR123.
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR123 (G protein-coupled receptor 123) is a 1,279 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 2 family and LN-TM7 subfamily. Existing as two alternatively spliced isoforms, GPR123 functions as an orphan receptor that is expressed in adult and fetal brain, and in adult spinal cord. The gene encoding GPR123 maps to human chromosome 10q26.3 and mouse chromosome 7 F4.Specifications
GPR123 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q86SQ6 | |
84435 | |
Peptide | |
Primary | |
-20°C | |
ADGRA1 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
G protein coupled receptor 123, GPR123, KIAA1828 | |
ADGRA1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title