Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR125 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575663
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575663 Supplier Thermo Scientific Supplier No. PA575663
Add to Cart
Add to Cart

GPR125 Polyclonal Antibody

GPR125 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR125
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ADGRA3
Gene Accession No. Q7TT36, Q8IWK6
Gene Alias 3830613O22Rik; Adgra2; ADGRA3; adhesion G protein-coupled receptor A2; adhesion G protein-coupled receptor A3; AU044632; G protein-coupled receptor 125; Gpr125; G-protein coupled receptor 125; PGR21; probable G-protein coupled receptor 125; TEM5L; Tem5-like; UNQ556/PRO1113
Gene Symbols ADGRA3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human GPR125.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166647, 305408, 70693
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.