Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR125 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120377 Supplier Invitrogen™ Supplier No. PA5120377
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse liver. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: ITAYLQCTRN THGSGIYPGN PQDERKAWRR CDRGGFWADD DYSRCQYAND VTRVLYMFNQ MPLNLTNAVA TARQLLAYTV EAANFSDKMD VIFVAEMIEK FGRFTKEEKS KELGDVMVDI ASNIMLADER VLWLAQREAK ACSRIVQCLQ RIATYRLAGG AHVYSTYSPN IALEAYVIKS TGFTGMTCTV FQKVAASDRT GLSDYGRRDP EGNLDKQLSF KCNVSNTFSS L.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR125 (G protein-coupled receptor 125), also known as PGR21 or TEM5L, is a 1,321 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 2 family and the LN-TM7 subfamily. Considered a novel orphan adhesion-type G-protein-coupled receptor, GPR125 has five leucine rich repeats (LRR), an immunoglobulin (Ig) domain and a GPS domain. GPR125 may play a functional role in choroidal and hippocampal response to brain injury. It is also suggested that GPR125 may be a marker for spermatogonial stem cells. Four isoforms of GPR125 exists due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR125
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ADGRA3
Gene Accession No. Q7TT36
Gene Alias 3830613O22Rik; Adgra2; ADGRA3; adhesion G protein-coupled receptor A2; adhesion G protein-coupled receptor A3; AU044632; G protein-coupled receptor 125; Gpr125; G-protein coupled receptor 125; PGR21; probable G-protein coupled receptor 125; TEM5L; Tem5-like; UNQ556/PRO1113
Gene Symbols ADGRA3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 370-600 of human ADGRA3 (NP_660333.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305408, 70693
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.