Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR126 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532795
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532795 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532795 Supplier Invitrogen™ Supplier No. PA532795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse (100%); Marmoset, Rat, Bovine, Dog, Panda, Rabbit, Pig (95%); Bat, Elephant, Horse (90%); Opossum, Platypus (80%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment with Proteinase K.

GPR126 encodes a product that is upregulated in human umbilical vein endothelial cells, encodes a G protein-coupled receptor. Variations in this gene can affect a persoNULLs stature. Multiple transcript variants encoding different proteins have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR126
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRG6
Gene Accession No. Q6F3F9, Q86SQ4
Gene Alias 1190004A11Rik; ADGRG6; ADGRG6 C-terminal fragment; ADGRG6 N-terminal fragment; ADGRG6-CTF; ADGRG6-NTF; adhesion G protein-coupled receptor G6; adhesion G-protein coupled receptor G6; AI449247; APG1; AW045736; bge; big ears; developmentally regulated G-protein-coupled receptor; DREG; G protein-coupled receptor 126; Gm222; gpr126; G-protein coupled receptor 126; HBV PreS1-transactivated protein 2; lau; lauscher; LCCS9; probable G-protein coupled receptor 126; PS1TP2; si:ch211-202p1.3; unm st49; unm_st49; vascular inducible G protein-coupled receptor; vascular-inducible G protein-coupled receptor; VIGR
Gene Symbols ADGRG6
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from N-Terminus of human GPR126.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215798, 57211
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.