Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR126 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550974
Description
GPR126 Polyclonal Antibody detects endogenous levels of GPR126 protein.
GPR126 encodes a product that is upregulated in human umbilical vein endothelial cells, encodes a G protein-coupled receptor. Variations in this gene can affect a persoNULLs stature. Multiple transcript variants encoding different proteins have been found for this gene.
Specifications
GPR126 | |
Polyclonal | |
Unconjugated | |
ADGRG6 | |
1190004A11Rik; ADGRG6; ADGRG6 C-terminal fragment; ADGRG6 N-terminal fragment; ADGRG6-CTF; ADGRG6-NTF; adhesion G protein-coupled receptor G6; adhesion G-protein coupled receptor G6; AI449247; APG1; AW045736; bge; big ears; developmentally regulated G-protein-coupled receptor; DREG; G protein-coupled receptor 126; Gm222; gpr126; G-protein coupled receptor 126; HBV PreS1-transactivated protein 2; lau; lauscher; LCCS9; probable G-protein coupled receptor 126; PS1TP2; si:ch211-202p1.3; unm st49; unm_st49; vascular inducible G protein-coupled receptor; vascular-inducible G protein-coupled receptor; VIGR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57211 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q86SQ4 | |
ADGRG6 | |
Synthesized peptide derived from the Internal region of human GPR126 (aa 431-480). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction