Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR126 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981882
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1882 Supplier Bioss Supplier No. BS12027R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR126 encodes a product that is upregulated in human umbilical vein endothelial cells, encodes a G protein-coupled receptor. Variations in this gene can affect a persoNULLs stature. Multiple transcript variants encoding different proteins have been found for this gene.

Specifications

Antigen GPR126
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADGRG6
Gene Accession No. Q86SQ4
Gene Alias ADGRG6; ADGRG6 C-terminal fragment; ADGRG6 N-terminal fragment; ADGRG6-CTF; ADGRG6-NTF; adhesion G protein-coupled receptor G6; Adhesion G-protein coupled receptor G6; APG1; bge; big ears; Developmentally regulated G-protein-coupled receptor; DREG; G protein-coupled receptor 126; GPR126; G-protein coupled receptor 126; HBV PreS1-transactivated protein 2; lau; lauscher; LCCS9; PS1TP2; si:ch211-202p1.3; unm st49; unm_st49; Vascular inducible G protein-coupled receptor; vascular-inducible G protein-coupled receptor; VIGR
Gene Symbols ADGRG6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human G-protein coupled receptor 126.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57211
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.