Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR128 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GPR128 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GPR128 Polyclonal specifically detects GPR128 in Human, Mouse samples. It is validated for Western Blot.Specifications
GPR128 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
FLJ14454, FLJ16382, FLJ29035, G protein-coupled receptor 128, MGC142011, MGC163260, probable G-protein coupled receptor 128 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 27-135 of human GPR128 (NP_116176.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS with 50% glycerol, pH7.3. | |
84873 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title