Learn More
GPR132 Polyclonal Antibody, Invitrogen™
Description
PA1-32755 detects GPCR G2A from human samples.
Specifications
Specifications
| Antigen | GPR132 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | GPR132 |
| Gene Accession No. | Q9UNW8 |
| Gene Alias | G protein-coupled receptor 132; G protein-coupled receptor G2A; G2 accumulation protein; G2a; GPR132; MGC99642; Probable G-protein coupled receptor 132 |
| Gene Symbols | GPR132 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.