Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR133 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533624
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533624 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533624 Supplier Invitrogen™ Supplier No. PA533624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Dog, Panda, Rabbit, Pig (100%) Gibbon, Bovine, Elephant, Horse (95%).

GPRs are characterized by the presence of 7 transmembrane domains. GPR133 is an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR133
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRD1
Gene Accession No. Q6QNK2
Gene Alias Adgrd1; adhesion G protein-coupled receptor D1; adhesion G-protein coupled receptor D1; DKFZp434B1272; E230012M21Rik; FLJ16770; G protein-coupled receptor 133; GPR133; G-protein coupled receptor 133; G-protein coupled receptor PGR25; MGC138512; MGC138514; Pgr25; probable G-protein coupled receptor 133
Gene Symbols ADGRD1
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human GPR133.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100352052, 100736874, 283383, 486229
Target Species Human, Canine, Rabbit, Monkey, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.