Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR137 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533626
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533626 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533626 Supplier Invitrogen™ Supplier No. PA533626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bat, Bovine, Elephant, Panda, Horse, Pig (100%) Hamster (81%).

GPR137 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR137
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR137
Gene Accession No. Q17QQ5, Q80ZU9, Q96N19
Gene Alias AI428855; C11orf4; G protein-coupled receptor 137; G protein-coupled receptor TM7SF1L1; GPR137; GPR137A; integral membrane protein GPR137; TM7SF1L1; transmembrane 7 superfamily member 1-like 1 protein
Gene Symbols GPR137
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 2nd cytoplasmic domain of human GPR137.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050142, 100524971, 107173, 505015, 56834, 612124, 689984
Target Species Human, Mouse, Rat, Canine, Monkey, Bovine, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.