Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR137 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569607
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
GPR137 is a protein coding gene.
Specifications
GPR137 | |
Polyclonal | |
Unconjugated | |
GPR137 | |
AI428855; C11orf4; G protein-coupled receptor 137; G protein-coupled receptor TM7SF1L1; GPR137; GPR137A; integral membrane protein GPR137; TM7SF1L1; transmembrane 7 superfamily member 1-like 1 protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
56834 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96N19 | |
GPR137 | |
synthetic peptide directed towards the N-terminal region of human GPR137. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction