Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR137A Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS16270R

Description
GPR137 is a protein coding gene.Specifications
GPR137A | |
Polyclonal | |
Unconjugated | |
GPR137 | |
AI428855; BOS_24962; C11orf4; G protein-coupled receptor 137; G protein-coupled receptor TM7SF1L1; GPR137; GPR137A; Integral membrane protein GPR137; Tm7sf1l1; Transmembrane 7 superfamily member 1-like 1 protein | |
Rabbit | |
Protein A | |
RUO | |
56834 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96N19 | |
GPR137 | |
KLH conjugated synthetic peptide derived from human GPR137A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction