Learn More
Invitrogen™ GPR137B Polyclonal Antibody

Description
PA1-32819 detects GPCR TM7SF1 from human samples.
Specifications
Specifications
| Antigen | GPR137B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | GPR137B |
| Gene Accession No. | O60478, Q8BNQ3 |
| Gene Alias | G protein-coupled receptor 137B; GPR137B; Integral membrane protein GPR137B; RP5-985L19.1; TM7SF1; transmembrane 7 superfamily member 1 (upregulated in kidney); transmembrane 7 superfamily member 1 protein |
| Gene Symbols | GPR137B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.