Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                GPR137B Rabbit anti-Human, Polyclonal, Proteintech
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | GPR137B | 
|---|---|
| Concentration | 0.17 mg/mL | 
| Applications | Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| 50-173-0988    | Proteintech Group Inc 164961AP150UL | 150 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each of 1 for $518.60
                                                 |  | |||||
Description
GPRs are characterized by the presence of 7 transmembrane domains. G Protein-Coupled Receptor (GPCR) TM7SF1 is a member of this family (subfamily: orphan U) and its expression has been reported in adult kidney and heart, but not in fetal kidney.Specifications
| GPR137B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O60478 | |
| 7107 | |
| GPR137B Fusion Protein Ag9633 | |
| Primary | |
| -20°C | |
| GPR137B | 
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GPR137B, TM7SF1 | |
| GPR137B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
			
             
                                                