Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR137B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110915
Description
Immunogen sequence: RVRNPTKDLT NPGMVPSHGF SPRSYFFDNP RRYDSDDDLA WNIAPQGLQG GFAPDYYDWG QQTNSFLAQA GTLQDSTLDP DKPSLG.
GPRs are characterized by the presence of 7 transmembrane domains. G Protein-Coupled Receptor (GPCR) TM7SF1 is a member of this family (subfamily: orphan U) and its expression has been reported in adult kidney and heart, but not in fetal kidney.
Specifications
GPR137B | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60478 | |
GPR137B | |
Recombinant Protein corresonding to Human GPR137B. Recombinant protein control fragment (Product #RP-89571). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
GPR137B | |
G protein-coupled receptor 137B; GPR137B; Integral membrane protein GPR137B; RP5-985L19.1; TM7SF1; transmembrane 7 superfamily member 1 (upregulated in kidney); transmembrane 7 superfamily member 1 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7107 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction