Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR137C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556554 Supplier Invitrogen™ Supplier No. PA556554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RAQRLNQNLA PAGMINSHSY SSRAYFFDNP RRYDSDDDLP RLGSSREGSL PNSQSLGWYG TMTGCGSSSY TVTPHLNGPM TDTAP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

GPR137C is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR137C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR137C
Gene Accession No. Q8N3F9
Gene Alias 6330416L11Rik; G protein-coupled receptor 137C; G protein-coupled receptor TM7SF1L2; Gm908; GPR137C; Integral membrane protein GPR137C; TM7SF1L2; Transmembrane 7 superfamily member 1-like 2 protein
Gene Symbols GPR137C
Host Species Rabbit
Immunogen Recombinant protein fragment of Human GPR137C (Product #RP-89102)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283554
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.