Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR137C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564894
Description
Immunogen sequence: RAQRLNQNLAP AGMINSHSYS SRAYFFDNPR RYDSDDDLPR LGSSREGSLP NSQSLGWYGT MTGCGSSSYT VTPHLNGPMT DTAP Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.
GPR137C is a protein coding gene.
Specifications
GPR137C | |
Polyclonal | |
Unconjugated | |
GPR137C | |
6330416L11Rik; G protein-coupled receptor 137C; G protein-coupled receptor TM7SF1L2; Gm908; GPR137C; Integral membrane protein GPR137C; TM7SF1L2; Transmembrane 7 superfamily member 1-like 2 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283554 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N3F9 | |
GPR137C | |
Recombinant Human GPR137C. Recombinant protein control fragment (Product #RP-89102). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction