Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR137C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102078 Supplier Invitrogen™ Supplier No. PA5102078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR137C.

GPR137C is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR137C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPR137C
Gene Accession No. Q8N3F9
Gene Alias 6330416L11Rik; G protein-coupled receptor 137C; G protein-coupled receptor TM7SF1L2; Gm908; GPR137C; Integral membrane protein GPR137C; TM7SF1L2; Transmembrane 7 superfamily member 1-like 2 protein
Gene Symbols GPR137C
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR137C(Accession Q8N3F9), corresponding to amino acid residues L256-V306.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283554, 70713
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.