Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR139 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533631
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533631 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533631 Supplier Invitrogen™ Supplier No. PA533631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR139 Polyclonal Antibody for IHC (P)

Orphan receptor that seems to act through a G(q/11)-mediated pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR139
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR139
Gene Accession No. P0C0W8, Q6DWJ6, Q80UC8
Gene Alias G protein-coupled receptor 139; G protein-coupled receptor PGR3; g(q)-coupled orphan receptor GPRg1; Gm495; Gpr139; GPRG1; G-protein coupled receptor 139; G-protein-coupled receptor PGR3; orphan G protein-coupled receptor TRHRL2; PGR3; Probable G-protein coupled receptor 139
Gene Symbols GPR139
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human GPR139.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100049812, 100353011, 124274, 209776, 293545, 431418, 489988, 540680
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Horse, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.