Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR141 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595714 Supplier Invitrogen™ Supplier No. PA595714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTTMAVINLV VVHSVFLLTV PFRLTYLIKK TWMFGLPFCK FVSAMLHIHM YLTFLFYVVI LVTRYLIFFK CKDKVEFYRK LHAVAASAGM WTLVIVIVVP L; Positive Samples: Mouse lung, Rat lung; Cellular Location: Cell membrane, Multi-pass membrane protein.

GPR141 is a member of the rhodopsin family of G protein-coupled receptors (GPRs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR141
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Gpr141
Gene Accession No. Q7TQP0
Gene Alias G protein-coupled receptor 141; GPR141; G-protein coupled receptor PGR13; Pgr13; probable G-protein coupled receptor 141
Gene Symbols Gpr141
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human GPR141 (NP_0013169221).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291179, 353346
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.