Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR144 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524067
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524067 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524067 Supplier Invitrogen™ Supplier No. PA524067
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR144 Polyclonal Antibody for Western Blot

GPR144 is an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR144
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADGRD2
Gene Accession No. Q7Z7M1
Gene Alias ADGRD2; adhesion G protein-coupled receptor D2; adhesion G-protein coupled receptor D2; G protein-coupled receptor 144; G protein-coupled receptor PGR24; GPR144; G-protein coupled receptor 144; G-protein coupled receptor GPR144; G-protein coupled receptor PGR24; PGR24
Gene Symbols ADGRD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 342-370 amino acids from the central region of human GPR144.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347088
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.