Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR144 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102087 Supplier Invitrogen™ Supplier No. PA5102087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR144.

GPR144 is an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR144
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRD2
Gene Accession No. Q7Z7M1
Gene Alias ADGRD2; adhesion G protein-coupled receptor D2; adhesion G-protein coupled receptor D2; G protein-coupled receptor 144; G protein-coupled receptor PGR24; GPR144; G-protein coupled receptor 144; G-protein coupled receptor GPR144; G-protein coupled receptor PGR24; PGR24
Gene Symbols ADGRD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADGRD2(Accession Q7Z7M1), corresponding to amino acid residues S314-R364.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347088
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.