Learn More
GPR146 Polyclonal Antibody, Invitrogen™
Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | GPR146 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7 |
| Gene | GPR146 |
| Gene Accession No. | Q96CH1 |
| Gene Alias | BC003323; G protein-coupled receptor 146; Gpr146; G-protein coupled receptor PGR8; PGR8; Probable G-protein coupled receptor 146; RGD1560731 |
| Gene Symbols | GPR146 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.