Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR147/NPFF1 Receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981874
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1874 Supplier Bioss Supplier No. BS12018R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The central nervous system neuropeptide FF (NPFF), is believed to play a role in pain modulation and opiate tolerance as well as other physiological functions. Two G protein-coupled receptors, NPFF1 and NPFF2, were isolated from the human central nervous system.

Specifications

Antigen GPR147/NPFF1 Receptor
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NPFFR1
Gene Accession No. Q9GZQ6
Gene Alias FLJ10751; G protein-coupled receptor 147; GPR147; G-protein coupled receptor 147; neuropeptide FF 1; Neuropeptide FF receptor 1; NPFF1; NPFF1 receptor; NPFF1R1; NPFFR1; NPFF-R1; OT7T022; RFamide-related peptide receptor OT7T022
Gene Symbols NPFFR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR147/NPFF1 Receptor .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64106
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.