Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR148 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533637
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Gibbon (93%).
Orphan receptor.
Specifications
| GPR148 | |
| Polyclonal | |
| Unconjugated | |
| GPR148 | |
| brain and testis restricted GPCR; BTR; G protein-coupled receptor 148; G protein-coupled receptor PGR6; GPR148; G-protein coupled receptor PGR6; PGR6; Probable G-protein coupled receptor 148 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 344561 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8TDV2 | |
| GPR148 | |
| Synthetic 14 amino acid peptide from C-terminus of human GPR148. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction