Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR148 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563869
Description
Immunogen sequence: LLRYRQLLGM VRGHLPSRRH QAIFTIS Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 41%.
Orphan receptor.
Specifications
GPR148 | |
Polyclonal | |
Unconjugated | |
GPR148 | |
brain and testis restricted GPCR; BTR; G protein-coupled receptor 148; G protein-coupled receptor PGR6; GPR148; G-protein coupled receptor PGR6; PGR6; Probable G-protein coupled receptor 148 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
344561 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDV2 | |
GPR148 | |
Recombinant protein corresponding to Human GPR148. Recombinant protein control fragment (Product #RP-95322). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction