Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533643
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Elephant, Pig (100%) Marmoset, Hamster (95%) Panda, Bat, Dog, Bovine, Rabbit, Horse (89%).
GPR15 expression has been reported in CD4(+) T lymphocytes, alveolar macrophages, and the basal surface of the small intestinal epithelium. ESTs have been isolated from normal brain and kidney cancer libraries.
Specifications
GPR15 | |
Polyclonal | |
Unconjugated | |
GPR15 | |
4933439K08Rik; BoB; brother of Bonzo; G protein-coupled receptor 15; GPR15; Gpr-15; G-protein coupled receptor 15; MGC126828; MGC126830 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100153787, 2838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P49685 | |
GPR15 | |
Synthetic 19 amino acid peptide from C-terminus of human GPR15. | |
50 μg | |
Primary | |
Human, Pig | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction