Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR150 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533645
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533645 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533645 Supplier Invitrogen™ Supplier No. PA533645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey (94%).

G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates, and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. G protein receptor 150 (GPR150), also designated PGR11 and seven transmembrane helix receptor, is a member of the rhodopsin family of GCPRs and is involved in signal transduction. GPR150 is expressed in various human tissues, including normal small intestine, skeletal muscle, kidney and tonsil, as well as cancerous blood, bladder, placenta and parathyroid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR150
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR150
Gene Accession No. Q8NGU9
Gene Alias C030001A19Rik; G protein-coupled receptor 150; Gpr150; G-protein coupled receptor PGR11; GPRv40; Pgr11; probable G-protein coupled receptor 150; RGD1564729; seven transmembrane helix receptor
Gene Symbols GPR150
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPR150.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285601
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.