Learn More
Invitrogen™ GPR150 Polyclonal Antibody

Description
Immunogen sequence: GDCRLRRQLR KRLGSLCCAP QGGAEDEEGP RGHQALYRQR WPHPHYHHAR REPLDEGGLR PPPPRPRPLP CSCESAF Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 69%.
Specifications
Specifications
| Antigen | GPR150 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GPR150 |
| Gene Accession No. | Q8NGU9 |
| Gene Alias | C030001A19Rik; G protein-coupled receptor 150; Gpr150; G-protein coupled receptor PGR11; GPRv40; Pgr11; probable G-protein coupled receptor 150; RGD1564729; seven transmembrane helix receptor |
| Gene Symbols | GPR150 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.