Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR151 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532803
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532803 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532803 Supplier Invitrogen™ Supplier No. PA532803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%); Gibbon (95%); Elephant (89%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

GPR151 is an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR151
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR151
Gene Accession No. Q8TDV0
Gene Alias AXOR40; C130082O03Rik; G protein-coupled receptor 151; galanin receptor 4; Galanin-receptor-like protein; GALR4; GalRL; GPCR; GPCR-2037; Gpr151; G-protein coupled receptor 151; G-protein coupled receptor 151 protein; G-protein coupled receptor PGR7; HGPRBMY7; nGPCR-2037; Pgr7; Probable G-protein coupled receptor 151; probable G-protein coupled receptor 151 protein; putative G protein-coupled receptor nGPCR-2037; putative G-protein coupled receptor
Gene Symbols GPR151
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 3rd extracellular domain of human GPR151.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134391
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.