Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR151 Polyclonal Antibody
GPR151 Polyclonal Antibody
Supplier: Thermo Scientific PA5102089
Description
GPR151 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
GPR151 | |
Polyclonal | |
Unconjugated | |
GPR151 | |
AXOR40; C130082O03Rik; G protein-coupled receptor 151; galanin receptor 4; Galanin-receptor-like protein; GALR4; GalRL; GPCR; GPCR-2037; Gpr151; G-protein coupled receptor 151; G-protein coupled receptor 151 protein; G-protein coupled receptor PGR7; HGPRBMY7; nGPCR-2037; Pgr7; Probable G-protein coupled receptor 151; probable G-protein coupled receptor 151 protein; putative G protein-coupled receptor nGPCR-2037; putative G-protein coupled receptor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
134391, 240239, 307475 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7TSN5, Q7TSN6, Q8TDV0 | |
GPR151 | |
A synthesized peptide derived from human GPR151(Accession Q8TDV0), corresponding to amino acid residues D385-K419. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction