Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR152 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532804
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532804 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532804 Supplier Invitrogen™ Supplier No. PA532804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Monkey (95%); Gibbon, Marmoset (89%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

G protein-coupled receptors (GPRs or GPCRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, are members of the largest protein family and play a role in many different stimulus-response pathways. G protein-coupled receptors mediate extracellular signals into intracellular signals (G protein activation). They respond to a wide variety of signaling molecules, including hormones, neurotransmitters and other proteins and peptides. GPR proteins are usually integral seven pass membrane proteins with some conserved amino acid regions. GPR-152 (G proteincoupled receptor 152) is a 511 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 family. Encoded by a gene on human chromosome 11q13.2 and mouse chromosome 19 A, GPR-152 may function as an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR152
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR152
Gene Accession No. Q8TDT2
Gene Alias A930009H15Rik; G protein-coupled receptor 152; G protein-coupled receptor PGR5; Gm673; Gpr152; G-protein coupled receptor PGR5; PGR5; Probable G-protein coupled receptor 152; putative G-protein coupled receptor
Gene Symbols GPR152
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human GPR152.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390212
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.