Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR152 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550975
Description
GPR152 Polyclonal Antibody detects endogenous levels of GPR152 protein.
G protein-coupled receptors (GPRs or GPCRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, are members of the largest protein family and play a role in many different stimulus-response pathways. G protein-coupled receptors mediate extracellular signals into intracellular signals (G protein activation). They respond to a wide variety of signaling molecules, including hormones, neurotransmitters and other proteins and peptides. GPR proteins are usually integral seven pass membrane proteins with some conserved amino acid regions. GPR-152 (G proteincoupled receptor 152) is a 511 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 family. Encoded by a gene on human chromosome 11q13.2 and mouse chromosome 19 A, GPR-152 may function as an orphan receptor.
Specifications
GPR152 | |
Polyclonal | |
Unconjugated | |
GPR152 | |
A930009H15Rik; G protein-coupled receptor 152; G protein-coupled receptor PGR5; Gm673; Gpr152; G-protein coupled receptor PGR5; PGR5; Probable G-protein coupled receptor 152; putative G-protein coupled receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
390212 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8TDT2 | |
GPR152 | |
Synthesized peptide derived from the N-terminal region of human GPR152. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction