Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR155 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150626B
Description
GPR155 Polyclonal specifically detects GPR155 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GPR155 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Q7Z3F1 | |
GPR155 | |
A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 800-900) [UniProt Q7Z3F1] | |
0.1 mL | |
Cancer, Neuroscience, Signal Transduction, Stem Cell Lines | |
151556 | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Biotin | |
PBS with 0.05% Sodium Azide | |
DEP.7, DEPDC3, FLJ31819, FLJ39346, G protein-coupled receptor 155, integral membrane protein GPR155, PGR22G-protein coupled receptor PGR22 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction