Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR155 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557547 Supplier Invitrogen™ Supplier No. PA557547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLIILPFKRR LEFLWNNKDT AENRDSPVSE EIKMTCQQFI HYHRDLCIRN IVKERRCGAK TSAGTFCGCD LVSW Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.

GPR155, an integral membrane protein related to G-protein coupled receptors, has been shown to have specific roles in Huntington disease and autism spectrum disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR155
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gpr155
Gene Accession No. Q7Z3F1
Gene Alias 1110017O10Rik; DEP.7; DEPDC3; F730029F15Rik; G protein-coupled receptor 155; GPR155; GPR155 variant 3; GPR155 variant 6; G-protein coupled receptor PGR22; integral membrane protein GPR155; PGR22
Gene Symbols Gpr155
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR155. Recombinant protein control fragment (Product #RP-96181).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151556
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.