Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR155 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983717
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3717 Supplier Bioss Supplier No. BS15380R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR155, an integral membrane protein related to G-protein coupled receptors, has been shown to have specific roles in Huntington disease and autism spectrum disorders.

Specifications

Antigen GPR155
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gpr155
Gene Accession No. Q7Z3F1
Gene Alias 1110017O10Rik; DEP.7; DEPDC3; F730029F15Rik; G protein-coupled receptor 155; GPR155; GPR155 variant 3; GPR155 variant 6; G-protein coupled receptor PGR22; Integral membrane protein GPR155; PGR22
Gene Symbols Gpr155
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR155.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151556
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.