Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR157 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983718
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3718 Supplier Bioss Supplier No. BS15381R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Orphan receptor that promotes neuronal differentiation of radial glial progenitors (RGPs). The activity of this receptor is mediated by a G(q)-protein that activates a phosphatidylinositol-calcium second messenger.

Specifications

Antigen GPR157
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPR157
Gene Accession No. Q5UAW9
Gene Alias F730108M23Rik; FLJ12132; G protein-coupled receptor 157; GPR157; G-protein coupled receptor 157; novel 7 transmembrane receptor (Secretin family) domain containing protein; probable G-protein coupled receptor 157
Gene Symbols GPR157
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR157.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80045
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.