Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GPR158 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | GPR158 |
---|---|
Clone | 1027651 |
Dilution | Flow Cytometry 0.25 ug/106 cells, Immunocytochemistry 8-25 ug/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB102861SP
![]() |
R&D Systems
MAB102861SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10286110
![]() |
R&D Systems
MAB102861100 |
100 μg |
Each for $432.50
|
|
|||||
Description
GPR158 Monoclonal antibody specifically detects GPR158 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
GPR158 | |
Flow Cytometry 0.25 ug/106 cells, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
G protein-coupled receptor 158, KIAA1136, KIAA1136FLJ37801, probable G-protein coupled receptor 158, RP11-59G22.1 | |
Human embryonic kidney cell HEK293-derived human GPR158 protein, Ala24-Gln411, Accession # Q5T848 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1027651 | |
Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q5T848 | |
57512 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title