Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR15L Antibody (1020904) [DyLight 550], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB104161L
Description
GPR15L Monoclonal antibody specifically detects GPR15L in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
GPR15L | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
387695 | |
Store at 4°C in the dark. | |
IgG2b |
ELISA, Flow Cytometry (Intracellular Staining), CyTOF | |
1020904 | |
ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
C10orf99 chromosome 10 open reading frame 99, UNQ1833 | |
E. coli-derived human C10orf99 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction