Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR160 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533650
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533650 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533650 Supplier Invitrogen™ Supplier No. PA533650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey, Marmoset (89%) Dog, Bovine (84%).

GPR160, part of the Class A family of G protein-coupled receptors (GPCRs), is prominently expressed in mouse embryonic stem cells and has been associated with various human solid tumors, particularly prostate cancer. The gene's expression contributes to the self-renewal and pluripotency of stem cells by participating in critical signaling pathways, including the JAK/STAT pathway, which is crucial for maintaining the stem cell cycle. High levels of GPR160 expression are observed in prostate cancer, though its role in disease progression is independent of the CARTp-mediated signaling pathway. Investigations into GPR160 have revealed its potential as a biomarker for prostate cancer. Additionally, it regulates mycobacteria entry into macrophages, implicating its functionality in immune responses. The gene's mechanisms of action, especially within cancer contexts, are subjects of ongoing research aimed at understanding its precise contributions to cellular proliferation and disease phenotypes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR160
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR160
Gene Accession No. Q9UJ42
Gene Alias 1700025D19Rik; G protein-coupled receptor 160; GPCR1; GPCR150; GPR160; G-protein coupled receptor GPCR1; hGPCR1; probable G-protein coupled receptor 160; putative G protein-coupled receptor
Gene Symbols GPR160
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 2nd extracellular domain of human GPR160.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.